Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133911
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
4
527
GenBank mRNA
BC062028.1
-
3
496
5623881
DN931898.1
-
3
545
FQ209537.1
-
4
527
FQ209603.1
-
4
527
FQ209620.1
-
3
512
FQ209757.1
-
4
528
FQ210253.1
-
4
527
FQ210412.1
-
4
527
FQ211152.1
-
4
527
FQ211263.1
-
4
526
FQ212554.1
-
4
527
FQ212743.1
-
4
527
FQ212875.1
-
4
527
FQ212927.1
-
4
528
FQ213085.1
-
4
527
FQ213427.1
-
4
527
FQ213510.1
-
4
527
FQ213847.1
-
4
528
FQ214087.1
-
4
527
FQ214354.1
-
4
527
FQ214515.1
-
4
527
FQ214553.1
-
4
528
FQ214850.1
-
4
527
FQ214997.1
-
4
527
FQ215007.1
-
4
527
FQ215548.1
-
4
529
FQ215662.1
-
4
527
FQ215682.1
-
4
527
FQ215870.1
-
4
527
FQ216120.1
-
4
528
FQ216296.1
-
4
527
FQ216389.1
-
4
528
FQ216406.1
-
4
530
FQ216495.1
-
4
527
FQ216618.1
-
4
527
FQ216655.1
-
4
528
FQ216733.1
-
4
529
FQ216774.1
-
4
527
FQ216813.1
-
4
526
FQ216851.1
-
4
527
FQ216913.1
-
4
528
FQ217406.1
-
4
527
FQ218561.1
-
4
527
FQ218832.1
-
4
527
FQ218943.1
-
4
527
FQ219025.1
-
4
530
FQ219184.1
-
4
531
FQ219389.1
-
4
527
FQ219457.1
-
4
527
FQ219678.1
-
4
527
FQ219815.1
-
4
527
FQ219817.1
-
4
527
FQ219837.1
-
4
527
FQ219855.1
-
4
527
FQ219923.1
-
4
527
FQ220008.1
-
4
527
FQ220051.1
-
4
527
FQ220077.1
-
4
527
FQ220115.1
-
4
527
FQ220181.1
-
4
527
FQ220201.1
-
4
525
FQ220213.1
-
4
527
FQ220247.1
-
4
528
FQ220282.1
-
4
529
FQ220284.1
-
4
527
FQ220302.1
-
4
527
FQ220338.1
-
4
527
FQ220345.1
-
4
527
FQ220370.1
-
4
527
FQ220406.1
-
4
527
FQ220413.1
-
4
527
FQ220420.1
-
4
528
FQ220433.1
-
4
524
FQ220444.1
-
4
527
FQ220445.1
-
4
523
FQ220449.1
-
4
527
FQ220500.1
-
4
527
FQ220514.1
-
4
527
FQ220527.1
-
4
527
FQ220533.1
-
4
559
FQ220542.1
-
4
527
FQ220545.1
-
4
527
FQ220634.1
-
4
527
FQ220654.1
-
4
527
FQ220684.1
-
4
528
FQ220690.1
-
4
529
FQ220769.1
-
4
527
FQ220787.1
-
4
527
FQ220801.1
-
4
527
FQ220833.1
-
4
527
FQ220838.1
-
4
527
FQ220888.1
-
4
527
FQ220932.1
-
4
528
FQ221146.1
-
4
527
FQ221380.1
-
4
527
FQ221533.1
-
4
528
FQ221556.1
-
4
525
FQ222171.1
-
4
527
FQ222181.1
-
4
527
FQ222327.1
-
4
527
FQ222772.1
-
4
527
FQ222906.1
-
4
527
FQ223121.1
-
4
527
FQ223487.1
-
4
527
FQ224547.1
-
4
527
FQ224834.1
-
4
527
FQ224846.1
-
4
527
FQ225170.1
-
4
527
FQ225608.1
-
4
527
FQ225654.1
-
4
529
FQ225860.1
-
4
529
FQ225886.1
-
4
527
FQ226306.1
-
4
529
FQ227381.1
-
4
527
FQ227522.1
-
4
522
FQ227719.1
-
4
527
FQ227837.1
-
4
527
FQ227937.1
-
4
528
FQ227941.1
-
4
527
FQ227942.1
-
4
647
FQ228548.1
-
4
527
FQ228995.1
-
4
527
FQ228996.1
-
4
527
FQ229139.1
-
4
529
FQ229159.1
-
4
527
FQ229164.1
-
4
528
FQ229208.1
-
4
527
FQ229241.1
-
4
527
FQ229244.1
-
4
527
FQ229248.1
-
2
407
FQ229272.1
-
4
527
FQ229273.1
-
4
528
FQ229303.1
-
4
527
FQ229337.1
-
4
527
FQ229352.1
-
4
527
FQ229365.1
-
4
527
FQ229397.1
-
4
527
FQ229408.1
-
4
528
FQ229433.1
-
4
527
FQ229448.1
-
4
527
FQ229521.1
-
4
527
FQ229547.1
-
4
527
FQ229554.1
-
4
527
FQ229566.1
-
4
523
FQ229610.1
-
4
527
FQ229626.1
-
4
524
FQ229726.1
-
4
527
FQ229740.1
-
4
527
FQ229742.1
-
4
527
FQ229776.1
-
4
527
FQ229808.1
-
4
527
FQ229815.1
-
4
527
FQ229822.1
-
4
527
FQ229830.1
-
4
529
FQ229883.1
-
4
529
FQ229901.1
-
4
527
FQ229929.1
-
4
527
FQ229932.1
-
2
425
FQ229952.1
-
4
527
FQ229974.1
-
4
527
FQ230005.1
-
4
527
FQ230055.1
-
4
526
FQ230127.1
-
4
527
FQ230267.1
-
4
530
FQ230943.1
-
4
527
FQ231445.1
-
4
529
FQ231525.1
-
4
527
FQ231591.1
-
4
529
FQ231710.1
-
4
528
FQ231733.1
-
4
528
FQ231897.1
-
4
529
FQ231967.1
-
4
527
FQ232029.1
-
4
528
FQ232154.1
-
4
527
FQ232165.1
-
4
527
FQ232310.1
-
4
526
FQ232319.1
-
4
528
FQ232697.1
-
4
529
FQ232767.1
-
4
527
FQ232951.1
-
4
527
FQ233152.1
-
4
526
FQ233377.1
-
4
527
FQ233473.1
-
4
527
FQ233732.1
-
4
527
FQ233808.1
-
4
527
FQ234195.1
-
4
527
FQ234341.1
-
4
527
FQ234613.1
-
4
527
FQ234733.1
-
4
527
FQ234735.1
-
4
417
FQ234863.1
-
4
527
FQ235063.1
-
4
527
FQ235079.1
-
4
527
FQ235122.1
-
4
527
FQ235259.1
-
4
527
X15096.1
-
4
527
Z29530.1
-
3
483
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-26xzd:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0