Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s133913
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
6
801
GenBank mRNA
BC062028.1
-
5
770
5623881
FQ209603.1
-
6
801
FQ209620.1
-
5
786
FQ209649.1
-
6
801
FQ209757.1
-
6
802
FQ210253.1
-
6
801
FQ210412.1
-
6
801
FQ211152.1
-
6
801
FQ211263.1
-
6
800
FQ212927.1
-
6
802
FQ213085.1
-
6
801
FQ213427.1
-
6
801
FQ213510.1
-
6
801
FQ213847.1
-
6
802
FQ214354.1
-
6
801
FQ214515.1
-
6
801
FQ214850.1
-
6
801
FQ214997.1
-
6
801
FQ215007.1
-
6
801
FQ215548.1
-
6
803
FQ215662.1
-
6
801
FQ215682.1
-
6
801
FQ215870.1
-
6
801
FQ216120.1
-
6
802
FQ216296.1
-
6
801
FQ216389.1
-
6
802
FQ216406.1
-
6
805
FQ216495.1
-
6
801
FQ216618.1
-
6
801
FQ216774.1
-
6
801
FQ216813.1
-
6
800
FQ216851.1
-
6
801
FQ216913.1
-
6
802
FQ217406.1
-
6
801
FQ218561.1
-
6
801
FQ218943.1
-
6
801
FQ219025.1
-
6
804
FQ219184.1
-
6
805
FQ219389.1
-
6
801
FQ219457.1
-
6
801
FQ219815.1
-
6
801
FQ219817.1
-
6
801
FQ219837.1
-
6
801
FQ219855.1
-
6
801
FQ219923.1
-
6
801
FQ220008.1
-
6
801
FQ220051.1
-
6
801
FQ220077.1
-
6
801
FQ220115.1
-
6
801
FQ220181.1
-
6
801
FQ220201.1
-
6
799
FQ220213.1
-
6
801
FQ220247.1
-
6
802
FQ220282.1
-
6
803
FQ220284.1
-
6
801
FQ220302.1
-
6
801
FQ220338.1
-
6
801
FQ220345.1
-
6
802
FQ220370.1
-
6
801
FQ220406.1
-
6
802
FQ220420.1
-
6
802
FQ220433.1
-
6
798
FQ220444.1
-
6
801
FQ220445.1
-
6
797
FQ220449.1
-
6
801
FQ220500.1
-
6
801
FQ220514.1
-
6
801
FQ220527.1
-
6
801
FQ220533.1
-
6
833
FQ220542.1
-
6
801
FQ220545.1
-
6
801
FQ220634.1
-
6
801
FQ220654.1
-
6
801
FQ220663.1
-
6
801
FQ220684.1
-
6
802
FQ220690.1
-
6
803
FQ220769.1
-
6
801
FQ220787.1
-
6
801
FQ220801.1
-
6
801
FQ220833.1
-
6
801
FQ220838.1
-
6
801
FQ220888.1
-
6
801
FQ220932.1
-
6
802
FQ221146.1
-
6
801
FQ221380.1
-
6
801
FQ221533.1
-
6
802
FQ221556.1
-
6
799
FQ222171.1
-
6
801
FQ222181.1
-
6
801
FQ222327.1
-
6
801
FQ222906.1
-
6
801
FQ223121.1
-
6
801
FQ223487.1
-
6
801
FQ224547.1
-
6
801
FQ224834.1
-
6
801
FQ224846.1
-
6
801
FQ225049.1
-
6
801
FQ225170.1
-
6
801
FQ225608.1
-
6
801
FQ225654.1
-
6
803
FQ225860.1
-
6
803
FQ225886.1
-
6
801
FQ226306.1
-
6
803
FQ227381.1
-
6
802
FQ227522.1
-
6
796
FQ227719.1
-
6
801
FQ227837.1
-
6
801
FQ227937.1
-
6
802
FQ227941.1
-
6
801
FQ227942.1
-
6
921
FQ228995.1
-
6
801
FQ228996.1
-
6
801
FQ229139.1
-
6
803
FQ229159.1
-
6
801
FQ229164.1
-
6
802
FQ229208.1
-
6
801
FQ229241.1
-
6
801
FQ229244.1
-
6
801
FQ229248.1
-
4
681
FQ229272.1
-
6
801
FQ229273.1
-
6
802
FQ229303.1
-
6
801
FQ229337.1
-
6
801
FQ229352.1
-
6
801
FQ229365.1
-
6
801
FQ229397.1
-
6
801
FQ229408.1
-
6
802
FQ229433.1
-
6
801
FQ229448.1
-
6
801
FQ229521.1
-
6
801
FQ229547.1
-
6
801
FQ229554.1
-
6
801
FQ229566.1
-
6
797
FQ229610.1
-
6
801
FQ229626.1
-
6
798
FQ229726.1
-
6
801
FQ229740.1
-
6
801
FQ229742.1
-
6
801
FQ229808.1
-
6
801
FQ229815.1
-
6
801
FQ229822.1
-
6
801
FQ229830.1
-
6
803
FQ229883.1
-
6
803
FQ229901.1
-
6
801
FQ229929.1
-
6
801
FQ229932.1
-
4
699
FQ229952.1
-
6
801
FQ229974.1
-
6
801
FQ230005.1
-
6
801
FQ230055.1
-
6
800
FQ230127.1
-
6
801
FQ230267.1
-
6
804
FQ231525.1
-
6
801
FQ231591.1
-
6
803
FQ231733.1
-
6
802
FQ231897.1
-
6
803
FQ231967.1
-
6
801
FQ232029.1
-
6
802
FQ232154.1
-
6
801
FQ232165.1
-
6
801
FQ232310.1
-
6
800
FQ232319.1
-
6
802
FQ232697.1
-
6
803
FQ232767.1
-
6
801
FQ232951.1
-
6
801
FQ233152.1
-
6
800
FQ233377.1
-
6
801
FQ233732.1
-
6
801
FQ233737.1
-
6
801
FQ233808.1
-
6
801
FQ234195.1
-
6
801
FQ234341.1
-
6
801
FQ234613.1
-
6
801
FQ234735.1
-
6
691
FQ234863.1
-
6
802
FQ235063.1
-
6
801
FQ235079.1
-
6
801
FQ235122.1
-
6
801
FQ235259.1
-
6
801
X15096.1
-
6
801
Z29530.1
-
5
757
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0