Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL12B SIRNA Assays ›
Gene Symbol:
Il12b
Gene Name:
interleukin 12B
Gene Aliases:
Il12
Chromosome Location:
Chr.10: 30034453 - 30048120 on Build Rnor_6.0
UniGene ID:
Rn.48686
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s134251
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il12b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022611.1
NP_072133.1
6
883
XM_006246147.2
XP_006246209.1
7
1092
GenBank mRNA
AF133197.1
-
6
883
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008352, AK162981, BC103608, M86671, BC103609, and others.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
natural killer cell activation involved in immune response
positive regulation of T-helper 1 type immune response
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
negative regulation of inflammatory response to antigenic stimulus
cell cycle arrest
cell surface receptor signaling pathway
response to UV-B
positive regulation of activation of JAK2 kinase activity
cell migration
cytokine-mediated signaling pathway
sensory perception of pain
natural killer cell activation
negative regulation of interleukin-10 production
negative regulation of interleukin-17 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell activation
positive regulation of natural killer cell proliferation
positive regulation of mononuclear cell proliferation
positive regulation of smooth muscle cell apoptotic process
T-helper cell differentiation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat4 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
defense response to protozoan
negative regulation of growth of symbiont in host
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
defense response to Gram-negative bacterium
positive regulation of NK T cell activation
positive regulation of NK T cell proliferation
defense response to virus
cellular response to lipopolysaccharide
cellular response to interferon-gamma
cytokine receptor activity
cytokine activity
cytokine receptor binding
growth factor activity
interleukin-12 alpha subunit binding
identical protein binding
protein homodimerization activity
interleukin-23 receptor binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline