Search
Search

SSTR2| Gene Symbol: SSTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001050.2 | NP_001041.1 | 2 | 471 | ||
| GenBank mRNA | AK290745.1 | - | 2 | 482 | ||
| AK313866.1 | - | 2 | 460 | |||
| BC009522.1 | - | 1 | 43 | |||
| BC019610.1 | - | 2 | 471 | 3875163 | ||
| BC095495.1 | - | 1 | 120 | 6971895 | ||
| BT019926.1 | - | 1 | 111 | |||
Set Membership: |
|