Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MSH2 SIRNA Assays ›
Gene Symbol:
Msh2
Gene Name:
mutS homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.6: 11215951 - 11274916 on Build Rnor_6.0
UniGene ID:
Rn.3174
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s135560
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Msh2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031058.1
NP_112320.1
7
1200
GenBank mRNA
BC161846.1
-
7
1216
9029023
X93591.1
-
7
1200
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
meiotic mismatch repair
in utero embryonic development
somatic recombination of immunoglobulin genes involved in immune response
oxidative phosphorylation
DNA repair
mismatch repair
postreplication repair
double-strand break repair
meiotic gene conversion
cellular response to DNA damage stimulus
cell cycle arrest
germ cell development
spermatogenesis
determination of adult lifespan
male gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
response to X-ray
response to UV-B
response to organic cyclic compound
somatic hypermutation of immunoglobulin genes
somatic recombination of immunoglobulin gene segments
B cell mediated immunity
B cell differentiation
intra-S DNA damage checkpoint
response to drug
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
response to amino acid
negative regulation of neuron apoptotic process
maintenance of DNA repeat elements
negative regulation of reciprocal meiotic recombination
isotype switching
negative regulation of DNA recombination
positive regulation of helicase activity
magnesium ion binding
four-way junction DNA binding
Y-form DNA binding
heteroduplex DNA loop binding
double-strand/single-strand DNA junction binding
DNA binding
damaged DNA binding
double-stranded DNA binding
single-stranded DNA binding
ATP binding
protein C-terminus binding
DNA-dependent ATPase activity
ATPase activity
centromeric DNA binding
enzyme binding
protein kinase binding
mismatched DNA binding
guanine/thymine mispair binding
single base insertion or deletion binding
dinucleotide insertion or deletion binding
single guanine insertion binding
single thymine insertion binding
dinucleotide repeat insertion binding
oxidized purine DNA binding
MutLalpha complex binding
protein homodimerization activity
ADP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0