Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STX1A SIRNA Assays ›
          Gene Symbol:
          STX1A
          Gene Name:
          syntaxin 1A
          Gene Aliases:
          HPC-1, P35-1, STX1, SYN1A
          Chromosome Location:
          Chr.7: 73699205 - 73719702 on Build GRCh38
          UniGene ID:
          Hs.647024
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13590

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STX1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165903.1 NP_001159375.1 4 293
          NM_004603.3 NP_004594.1 4 293
          XM_017012567.1 XP_016868056.1 4 308
          GenBank mRNA AB086954.1 - 4 237
          AK313980.1 - 4 241
          BC003011.1 - 4 265 2820851
          BC064644.1 - 4 264 5769528
          CR457018.1 - 4 237
          D37932.1 - 4 237
          DA159446.1 - 4 293
          KJ534952.1 - 4 237
          KJ534980.1 - 4 237
          L37792.1 - 4 238
          U12918.1 - 3 196

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of neurotransmitter secretion
          intracellular protein transport
          neurotransmitter secretion
          response to gravity
          positive regulation of norepinephrine secretion
          regulation of synaptic vesicle priming
          glutamate secretion
          synaptic vesicle docking
          synaptic vesicle fusion to presynaptic active zone membrane
          secretion by cell
          positive regulation of catecholamine secretion
          positive regulation of calcium ion-dependent exocytosis
          vesicle docking
          regulation of insulin secretion
          positive regulation of excitatory postsynaptic potential
          SNARE binding
          glycoprotein binding
          SNAP receptor activity
          protein binding
          myosin binding
          calcium channel inhibitor activity
          chloride channel inhibitor activity
          kinase binding
          protein domain specific binding
          protein binding, bridging
          myosin head/neck binding
          ATP-dependent protein binding
          ion channel binding
          protein heterodimerization activity
          protein N-terminus binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0