Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SFRP1 SIRNA Assays ›
Gene Symbol:
Sfrp1
Gene Name:
secreted frizzled-related protein 1
Gene Aliases:
sFRP-1
Chromosome Location:
Chr.16: 73372007 - 73410777 on Build Rnor_6.0
UniGene ID:
Rn.163333
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s136553
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sfrp1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276712.1
NP_001263641.1
2, 3
625
GenBank mRNA
AF167308.1
-
2, 3
221
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
ureteric bud development
somitogenesis
neural tube closure
hematopoietic progenitor cell differentiation
proteolysis
multicellular organism development
positive regulation of cell proliferation
negative regulation of cell proliferation
male gonad development
female gonad development
cellular response to starvation
dorsal/ventral axis specification
anterior/posterior pattern specification
regulation of cell cycle process
negative regulation of gene expression
negative regulation of epithelial to mesenchymal transition
neural crest cell fate commitment
response to organic cyclic compound
neural tube development
hemopoiesis
positive regulation of Wnt signaling pathway
negative regulation of Wnt signaling pathway
regulation of ossification
negative regulation of ossification
positive regulation of cell growth
negative regulation of cell growth
negative regulation of cell migration
negative regulation of BMP signaling pathway
negative regulation of osteoblast proliferation
somatic stem cell population maintenance
non-canonical Wnt signaling pathway
response to drug
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of JUN kinase activity
cellular response to fibroblast growth factor stimulus
stromal-epithelial cell signaling involved in prostate gland development
negative regulation of B cell differentiation
positive regulation of fat cell differentiation
negative regulation of osteoblast differentiation
negative regulation of osteoclast differentiation
positive regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
development of primary male sexual characteristics
negative regulation of insulin secretion
negative regulation of bone remodeling
negative regulation of fibroblast proliferation
digestive tract morphogenesis
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
negative regulation of peptidyl-tyrosine phosphorylation
canonical Wnt signaling pathway
Wnt signaling pathway, planar cell polarity pathway
hematopoietic stem cell differentiation
bone trabecula formation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
regulation of branching involved in prostate gland morphogenesis
negative regulation of androgen receptor signaling pathway
cellular response to vitamin D
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
cellular response to prostaglandin E stimulus
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to hypoxia
cellular response to X-ray
cellular response to heparin
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
negative regulation of canonical Wnt signaling pathway
regulation of establishment of planar polarity
planar cell polarity pathway involved in neural tube closure
Wnt signaling pathway involved in somitogenesis
convergent extension involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of planar cell polarity pathway involved in axis elongation
positive regulation of non-canonical Wnt signaling pathway
negative regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
negative regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
negative regulation of fibroblast apoptotic process
positive regulation of fibroblast apoptotic process
positive regulation of extrinsic apoptotic signaling pathway
molecular_function
cysteine-type endopeptidase activity
G-protein coupled receptor activity
frizzled binding
drug binding
heparin binding
Wnt-protein binding
identical protein binding
Wnt-activated receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0