Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SVIL SIRNA Assays ›
          Gene Symbol:
          SVIL SVILP1
          Gene Name:
          supervillin
          supervillin pseudogene 1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.10: 29457338 - 29737001 on Build GRCh38
          Chromosome Location:
          Chr.10: 30692274 - 30717013 on Build GRCh38
          UniGene ID:
          Hs.499209
          Hs.408581
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s13662

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SVIL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001323599.1 NP_001310528.1 28 4724
          NM_001323600.1 NP_001310529.1 26 4325
          NM_003174.3 NP_003165.2 25 4110
          NM_021738.2 NP_068506.2 27 5691
          XM_005252570.2 XP_005252627.1 31 5714
          XM_005252571.3 XP_005252628.1 31 5640
          XM_005252573.2 XP_005252630.1 29 5846
          XM_011519630.2 XP_011517932.2 28 6481
          XM_011519631.2 XP_011517933.2 28 6463
          XM_011519632.2 XP_011517934.2 28 6388
          XM_011519633.2 XP_011517935.2 26 6337
          XM_011519634.1 XP_011517936.1 31 5772
          XM_011519635.1 XP_011517937.1 29 5739
          XM_011519636.2 XP_011517938.2 28 5629
          XM_011519637.2 XP_011517939.2 26 5377
          XM_011519638.2 XP_011517940.2 25 5281
          XM_017016575.1 XP_016872064.1 29 6559
          XM_017016576.1 XP_016872065.1 28 6478
          XM_017016577.1 XP_016872066.1 27 6433
          XM_017016578.1 XP_016872067.1 27 5533
          XM_017016579.1 XP_016872068.1 26 5503
          XM_017016580.1 XP_016872069.1 27 5458
          XM_017016581.1 XP_016872070.1 24 5251
          XM_017016582.1 XP_016872071.1 23 5155
          GenBank mRNA AF051850.1 - 25 4110
          AF051851.1 - 21 3760
          AF109135.1 - 27 5691
          AK302077.1 - 10 1396
          AK304720.1 - 13 1716
          AK304799.1 - 11 1474
          AK310272.1 - 10 1357
          AL117410.1 - 12 1768
          JX467682.1 - 22 4842
          Gene Symbol: SVILP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036438.1 5 875
          GenBank mRNA AK302694.1 - 5 875

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          skeletal muscle tissue development
          positive regulation of cytokinesis
          protein binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0