Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYK SIRNA Assays ›
          Gene Symbol:
          SYK
          Gene Name:
          spleen associated tyrosine kinase
          Gene Aliases:
          p72-Syk
          Chromosome Location:
          Chr.9: 90801680 - 90898560 on Build GRCh38
          UniGene ID:
          Hs.371720
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13681

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135052.3 NP_001128524.1 4 905
          NM_001174167.2 NP_001167638.1 4 851
          NM_001174168.2 NP_001167639.1 4 770
          NM_003177.6 NP_003168.2 4 905
          XM_005252147.3 XP_005252204.1 4 816
          XM_011518946.2 XP_011517248.1 4 765
          GenBank mRNA AK290927.1 - 4 749
          BC001645.2 - 4 797 2959066
          BC002962.1 - 4 797 3542895
          BC011399.1 - 4 754 3870426
          FN391891.1 - 3 650
          FN391892.1 - 3 650
          FN391893.1 - 3 650
          L28824.1 - 4 797
          S80267.1 - 3 651
          X73568.1 - 4 789
          Z29630.1 - 4 798

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          lymph vessel development
          positive regulation of receptor internalization
          stimulatory C-type lectin receptor signaling pathway
          adaptive immune response
          macrophage activation involved in immune response
          neutrophil activation involved in immune response
          leukocyte activation involved in immune response
          serotonin secretion by platelet
          protein phosphorylation
          inflammatory response
          leukocyte cell-cell adhesion
          transmembrane receptor protein tyrosine kinase signaling pathway
          integrin-mediated signaling pathway
          activation of JUN kinase activity
          cell proliferation
          organ morphogenesis
          regulation of platelet activation
          regulation of tumor necrosis factor-mediated signaling pathway
          viral process
          peptidyl-serine phosphorylation
          leukotriene biosynthetic process
          platelet activation
          neutrophil chemotaxis
          receptor internalization
          positive regulation of type I interferon production
          regulation of superoxide anion generation
          positive regulation of cell adhesion mediated by integrin
          peptidyl-tyrosine autophosphorylation
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          defense response to bacterium
          transcription factor import into nucleus
          positive regulation of mast cell degranulation
          regulation of neutrophil degranulation
          beta selection
          innate immune response
          positive regulation of interleukin-3 biosynthetic process
          positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of bone resorption
          positive regulation of alpha-beta T cell differentiation
          positive regulation of alpha-beta T cell proliferation
          blood vessel morphogenesis
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of phagocytosis
          positive regulation of calcium-mediated signaling
          B cell receptor signaling pathway
          regulation of sequence-specific DNA binding transcription factor activity
          regulation of ERK1 and ERK2 cascade
          cellular response to molecule of fungal origin
          cellular response to low-density lipoprotein particle stimulus
          regulation of arachidonic acid secretion
          regulation of platelet aggregation
          protein kinase activity
          protein serine/threonine kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          integrin binding
          protein binding
          ATP binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0