Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYT1 SIRNA Assays ›
          Gene Symbol:
          SYT1
          Gene Name:
          synaptotagmin 1
          Gene Aliases:
          P65, SVP65, SYT
          Chromosome Location:
          Chr.12: 78863993 - 79452008 on Build GRCh38
          UniGene ID:
          Hs.310545
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13694

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135805.1 NP_001129277.1 8, 9 1306
          NM_001135806.1 NP_001129278.1 6, 7 892
          NM_001291901.1 NP_001278830.1 6, 7 1217
          NM_005639.2 NP_005630.1 7, 8 1292
          XM_005269113.2 XP_005269170.1 7, 8 1282
          XM_006719576.1 XP_006719639.1 6, 7 905
          XM_011538710.1 XP_011537012.1 6, 7 1225
          XM_017019909.1 XP_016875398.1 6, 7 885
          GenBank mRNA AK094616.1 - 8, 9 1306
          AK126908.1 - 7, 8 1292
          AK308743.1 - 4, 5 408
          BC058917.1 - 7, 8 900 6187902
          BX647240.1 - 7, 8 1186
          CR627387.1 - 6, 7 892
          M55047.1 - 4, 5 662

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of calcium ion
          vesicle fusion
          chemical synaptic transmission
          neurotransmitter secretion
          brain development
          glutamate secretion
          regulation of exocytosis
          regulation of calcium ion-dependent exocytosis
          positive regulation of vesicle fusion
          positive regulation of calcium ion-dependent exocytosis
          vesicle docking
          synaptic vesicle endocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          positive regulation of synaptic transmission
          protein homooligomerization
          regulation of synaptic transmission, glutamatergic
          cellular response to calcium ion
          fast, calcium ion-dependent exocytosis of neurotransmitter
          regulation of regulated secretory pathway
          positive regulation of dendrite extension
          SNARE binding
          phosphatidylserine binding
          calcium ion binding
          protein binding
          calmodulin binding
          calcium-dependent phospholipid binding
          1-phosphatidylinositol binding
          phosphatidylinositol-4,5-bisphosphate binding
          protein C-terminus binding
          syntaxin-1 binding
          clathrin binding
          identical protein binding
          calcium-dependent protein binding
          low-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0