Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HNF1B SIRNA Assays ›
          Gene Symbol:
          HNF1B
          Gene Name:
          HNF1 homeobox B
          Gene Aliases:
          FJHN, HNF-1-beta, HNF-1B, HNF1beta, HNF2, HPC11, LF-B3, LFB3, MODY5, TCF-2, TCF2, VHNF1
          Chromosome Location:
          Chr.17: 37686431 - 37745078 on Build GRCh38
          UniGene ID:
          Hs.191144
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s13871

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HNF1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000458.3 NP_000449.1 4 1151
          NM_001165923.3 NP_001159395.1 4 1073
          NM_001304286.1 NP_001291215.1 4 1073
          XM_011525160.1 XP_011523462.1 4 1132
          XM_011525161.1 XP_011523463.1 4 1132
          XM_011525162.2 XP_011523464.1 4 1132
          XM_011525163.2 XP_011523465.1 4 1132
          XM_011525164.1 XP_011523466.1 4 1054
          GenBank mRNA AK290131.1 - 4 1132
          AK296633.1 - 4 1054
          AK298393.1 - 5 1001
          BC017714.1 - 4 1145 4414015
          BT007126.1 - 4 957
          CR536572.1 - 4 957
          HM116553.1 - 4 1013
          HM116554.1 - 4 994
          HM116555.1 - 4 1091
          HM116556.1 - 4 1013
          X58840.1 - 4 1151
          X71348.1 - 2 409

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          endodermal cell fate specification
          kidney development
          inner cell mass cell differentiation
          transcription, DNA-templated
          Notch signaling pathway
          response to glucose
          anterior/posterior pattern specification
          response to organic cyclic compound
          insulin secretion
          regulation of Wnt signaling pathway
          hindbrain development
          circadian regulation of gene expression
          regulation of pronephros size
          pronephric nephron tubule development
          response to drug
          regulation of endodermal cell fate specification
          positive regulation of transcription, DNA-templated
          embryonic digestive tract morphogenesis
          branching morphogenesis of an epithelial tube
          pronephros development
          epithelial cell proliferation
          positive regulation of transcription initiation from RNA polymerase II promoter
          ureteric bud elongation
          hepatoblast differentiation
          negative regulation of mesenchymal cell apoptotic process involved in mesonephric nephron morphogenesis
          protein-DNA complex assembly
          hepatocyte differentiation
          regulation of branch elongation involved in ureteric bud branching
          mesonephric duct formation
          negative regulation of mesenchymal cell apoptotic process involved in metanephros development
          RNA polymerase II regulatory region sequence-specific DNA binding
          core promoter proximal region DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          protein complex binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0