Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ILK SIRNA Assays ›
Gene Symbol:
Ilk
Gene Name:
integrin-linked kinase
Gene Aliases:
-
Chromosome Location:
Chr.1: 170578941 - 170585192 on Build Rnor_6.0
UniGene ID:
Rn.95042
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s139498
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ilk
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_133409.2
NP_596900.1
2, 3
321
GenBank mRNA
AF329194.1
-
2, 3
258
BC062406.1
-
2, 3
321
5623251
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010562, NM_001161724, AK154207, AK205321, AK213330, and others.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
branching involved in ureteric bud morphogenesis
positive regulation of cell-matrix adhesion
protein phosphorylation
negative regulation of protein kinase activity
cell cycle arrest
integrin-mediated signaling pathway
cell aging
positive regulation of cell proliferation
negative regulation of cardiac muscle cell apoptotic process
fibroblast migration
Schwann cell development
negative regulation of smooth muscle cell migration
peptidyl-serine phosphorylation
nerve development
myelination in peripheral nervous system
cell projection organization
positive regulation of cell migration
positive regulation of BMP signaling pathway
myelin assembly
regulation of actin cytoskeleton organization
substrate adhesion-dependent cell spreading
positive regulation of phosphorylation
negative regulation of apoptotic process
extracellular fibril organization
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
protein kinase B signaling
negative regulation of neuron apoptotic process
establishment or maintenance of epithelial cell apical/basal polarity
positive regulation of myoblast differentiation
positive regulation of osteoblast differentiation
positive regulation of axon extension
positive regulation of transcription, DNA-templated
negative regulation of smooth muscle cell proliferation
neuron projection morphogenesis
positive regulation of axonogenesis
positive regulation of dendrite morphogenesis
protein heterooligomerization
positive regulation of protein kinase B signaling
platelet aggregation
positive regulation of canonical Wnt signaling pathway
negative regulation of neural precursor cell proliferation
protein kinase activity
protein serine/threonine kinase activity
signal transducer activity
integrin binding
protein binding
ATP binding
SH3 domain binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0