Search
Search

Dkc1| Gene Symbol: Dkc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133419.1 | NP_596910.1 | 7 | 703 | ||
| XM_008773596.1 | XP_008771818.1 | 7 | 1149 | |||
| GenBank mRNA | BC087600.1 | - | 7 | 662 | ||
| BC099832.1 | - | 7 | 661 | 7934021 | ||
| Z34922.1 | - | 7 | 703 | |||