Search
Search

Cd40| Gene Symbol: Cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134360.1 | NP_599187.1 | 9 | 743 | ||
| XM_006235511.2 | XP_006235573.1 | 10 | 865 | |||
| XM_008762463.1 | XP_008760685.1 | 9 | 799 | |||
| XM_008762464.1 | XP_008760686.1 | 9 | 799 | |||
| XM_008762465.1 | XP_008760687.1 | 8 | 743 | |||
| GenBank mRNA | BC097949.1 | - | 9 | 743 | 7387440 | |
| FQ226992.1 | - | 10 | 786 | |||