Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APAF1 SIRNA Assays ›
          Gene Symbol:
          APAF1
          Gene Name:
          apoptotic peptidase activating factor 1
          Gene Aliases:
          APAF-1, CED4
          Chromosome Location:
          Chr.12: 98645141 - 98735433 on Build GRCh38
          UniGene ID:
          Hs.552567
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1415

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APAF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001160.2 NP_001151.1 4 966
          NM_013229.2 NP_037361.1 4 966
          NM_181861.1 NP_863651.1 4 999
          NM_181868.1 NP_863658.1 4 999
          NM_181869.1 NP_863659.1 4 999
          XM_017019250.1 XP_016874739.1 4 1158
          GenBank mRNA AB007873.2 - 3 389
          AB103079.1 - 3 467
          AF013263.1 - 4 966
          AF134397.1 - 3 389
          AF149794.1 - 3 422
          AJ243003.1 - 3 422
          AJ243004.1 - 3 422
          AJ243005.1 - 3 422
          AJ243006.1 - 3 422
          AJ243007.1 - 3 422
          AJ243008.1 - 3 422
          AJ243009.1 - 3 422
          AJ243010.1 - 3 422
          AJ243011.1 - 3 422
          AJ243048.1 - 3 422
          AJ243107.1 - 3 422
          AK307509.1 - 4 1158
          BC136531.1 - 4 570 9020521
          BC136532.1 - 4 570 9020522
          BC150255.1 - 3 418 9007183
          EF560718.1 - 3 427

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          neural tube closure
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          nervous system development
          aging
          response to nutrient
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          cardiac muscle cell apoptotic process
          cell differentiation
          forebrain development
          glial cell apoptotic process
          regulation of apoptotic process
          positive regulation of apoptotic process
          protein homooligomerization
          neuron apoptotic process
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to transforming growth factor beta stimulus
          response to G1 DNA damage checkpoint signaling
          intrinsic apoptotic signaling pathway
          regulation of apoptotic DNA fragmentation
          positive regulation of apoptotic signaling pathway
          nucleotide binding
          protein binding
          ATP binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          heat shock protein binding
          identical protein binding
          ADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jdrcd:80/100.66.76.32:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0