Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJP1 SIRNA Assays ›
Gene Symbol:
TJP1
Gene Name:
tight junction protein 1
Gene Aliases:
ZO-1
Chromosome Location:
Chr.15: 29699367 - 29968919 on Build GRCh38
UniGene ID:
Hs.743990
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14157
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301025.1
NP_001287954.1
24
4322
NM_001301026.1
NP_001287955.1
23
4097
NM_003257.4
NP_003248.3
23
4558
NM_175610.3
NP_783297.2
22
4318
XM_005254617.3
XP_005254674.2
24
4442
XM_005254618.3
XP_005254675.1
23
4189
XM_005254619.3
XP_005254676.2
23
4202
XM_005254620.3
XP_005254677.2
23
4202
XM_011521972.2
XP_011520274.2
24
4442
XM_017022521.1
XP_016878010.1
24
4442
XM_017022522.1
XP_016878011.1
24
4442
XM_017022523.1
XP_016878012.1
24
4442
XM_017022524.1
XP_016878013.1
23
4202
XM_017022525.1
XP_016878014.1
23
4204
XM_017022526.1
XP_016878015.1
23
4189
XM_017022527.1
XP_016878016.1
23
4230
XM_017022528.1
XP_016878017.1
23
4204
XM_017022529.1
XP_016878018.1
23
4189
XM_017022530.1
XP_016878019.1
22
3949
XM_017022531.1
XP_016878020.1
24
4272
GenBank mRNA
AB370197.1
-
22
4062
AK302971.1
-
4
1123
AK304758.1
-
23
4189
BC111712.1
-
23
4460
40037646
BX640879.1
-
23
4098
CR749235.1
-
15
2851
DA752892.1
-
3
231
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
blastocyst formation
cell-cell junction assembly
sensory perception of sound
hippo signaling
establishment of endothelial intestinal barrier
cell-cell adhesion
cell-cell signaling involved in cell-cell junction organization
protein binding
calmodulin binding
protein domain specific binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0