Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBA2 SIRNA Assays ›
Gene Symbol:
APBA2
Gene Name:
amyloid beta precursor protein binding family A member 2
Gene Aliases:
D15S1518E, HsT16821, LIN-10, MGC:14091, MINT2, X11-BETA, X11L
Chromosome Location:
Chr.15: 28885479 - 29118315 on Build GRCh38
UniGene ID:
Hs.618112
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1425
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBA2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130414.1
NP_001123886.1
8
1546
NM_005503.3
NP_005494.2
9
1582
XM_011521488.2
XP_011519790.1
10
1718
XM_011521489.2
XP_011519791.1
11
2569
XM_011521490.2
XP_011519792.1
11
2538
XM_011521491.1
XP_011519793.1
9
1597
XM_011521492.2
XP_011519794.1
9
1847
XM_011521493.2
XP_011519795.1
9
2349
XM_011521494.2
XP_011519796.1
10
2379
XM_011521495.1
XP_011519797.1
7
866
XM_011521496.1
XP_011519798.1
6
830
XM_017022101.1
XP_016877590.1
10
2379
XM_017022102.1
XP_016877591.1
11
2469
XM_017022103.1
XP_016877592.1
11
1788
XM_017022104.1
XP_016877593.1
10
1811
XM_017022105.1
XP_016877594.1
10
1764
XM_017022106.1
XP_016877595.1
10
2375
XM_017022107.1
XP_016877596.1
10
1844
XM_017022108.1
XP_016877597.1
10
2344
XM_017022109.1
XP_016877598.1
11
2465
XM_017022110.1
XP_016877599.1
11
2565
XM_017022111.1
XP_016877600.1
9
1708
XM_017022112.1
XP_016877601.1
10
1761
XM_017022113.1
XP_016877602.1
9
2343
XM_017022114.1
XP_016877603.1
9
2312
XM_017022115.1
XP_016877604.1
8
1672
XM_017022116.1
XP_016877605.1
8
1561
XM_017022117.1
XP_016877606.1
9
1901
GenBank mRNA
AB014719.1
-
7
1396
AB209281.1
-
8
1453
AF047348.1
-
9
1573
AK124794.1
-
6
830
AK310413.1
-
9
1662
BC082986.1
-
8
1525
6671740
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
chemical synaptic transmission
nervous system development
locomotory behavior
regulation of gene expression
protein transport
multicellular organism growth
beta-amyloid binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0