Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPM1 SIRNA Assays ›
          Gene Symbol:
          TPM1
          Gene Name:
          tropomyosin 1 (alpha)
          Gene Aliases:
          C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
          Chromosome Location:
          Chr.15: 63042196 - 63071915 on Build GRCh38
          UniGene ID:
          Hs.133892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14340
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000366.5 NP_000357.3 8 924
          NM_001018004.1 NP_001018004.1 8 924
          NM_001018005.1 NP_001018005.1 8 924
          NM_001018006.1 NP_001018006.1 8 924
          NM_001018007.1 NP_001018007.1 8 924
          NM_001018008.1 NP_001018008.1 7 764
          NM_001018020.1 NP_001018020.1 8 924
          NM_001301244.1 NP_001288173.1 8 924
          NM_001301289.1 NP_001288218.1 7 764
          XM_005254637.1 XP_005254694.1 9 1005
          XM_005254638.3 XP_005254695.2 8 1367
          XM_005254639.3 XP_005254696.2 8 1367
          XM_005254645.2 XP_005254702.2 8 1367
          XM_005254646.2 XP_005254703.1 7 750
          XM_005254647.3 XP_005254704.1 7 750
          XM_005254650.3 XP_005254707.1 7 750
          XM_005254651.1 XP_005254708.1 7 750
          XM_005254652.1 XP_005254709.1 7 750
          XM_005254653.2 XP_005254710.1 7 750
          XM_006720667.3 XP_006720730.2 8 1367
          XM_017022534.1 XP_016878023.1 8 1367
          XM_017022535.1 XP_016878024.1 8 879
          XM_017022536.1 XP_016878025.1 8 1367
          XM_017022537.1 XP_016878026.1 8 879
          XM_017022538.1 XP_016878027.1 8 1367
          XM_017022539.1 XP_016878028.1 8 1367
          XM_017022540.1 XP_016878029.1 7 750
          XM_017022541.1 XP_016878030.1 7 750
          GenBank mRNA AB209041.1 - 7 801
          AF474156.1 - 7 526
          AJ000147.1 - 1 25
          AJ001055.1 - 1 25
          AK057989.1 - 10 710
          AK092051.1 - 6 2582
          AK092573.1 - 7 1822
          AK129882.1 - 7 611
          AK131384.1 - 9 938
          AK298625.1 - 7 640
          AK299387.1 - 8 813
          AK301319.1 - 7 660
          AL050179.1 - 8 805
          AY640414.1 - 8 733
          AY640415.1 - 8 733
          BC007433.2 - 8 874 2962925
          BC050473.1 - 7 751
          BC053545.1 - 7 762 5730451
          BX458841.2 - 8 793
          CD679271.1 - 4 255
          DQ424903.1 - 7 625
          GQ472198.1 - 8 874
          KU178366.1 - 8 733
          KU178367.1 - 8 733
          M19713.1 - 8 789
          M19714.1 - 8 675
          M19715.1 - 7 570
          X12369.1 - 7 563
          Z24727.1 - 948

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00165966_m1

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of heart rate by epinephrine
          movement of cell or subcellular component
          muscle contraction
          regulation of muscle contraction
          cytoskeleton organization
          regulation of heart contraction
          regulation of cell shape
          muscle filament sliding
          negative regulation of cell migration
          ruffle organization
          positive regulation of ATPase activity
          cellular response to reactive oxygen species
          wound healing
          sarcomere organization
          positive regulation of cell adhesion
          positive regulation of stress fiber assembly
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          negative regulation of vascular smooth muscle cell proliferation
          negative regulation of vascular associated smooth muscle cell migration
          actin binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          structural constituent of muscle

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0