Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPM1 SIRNA Assays ›
          Gene Symbol:
          TPM1
          Gene Name:
          tropomyosin 1 (alpha)
          Gene Aliases:
          C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
          Chromosome Location:
          Chr.15: 63042196 - 63071915 on Build GRCh38
          UniGene ID:
          Hs.133892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14341
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000366.5 NP_000357.3 8 954
          NM_001018004.1 NP_001018004.1 8 954
          NM_001018005.1 NP_001018005.1 8 954
          NM_001018006.1 NP_001018006.1 8 954
          NM_001018007.1 NP_001018007.1 8 954
          NM_001018008.1 NP_001018008.1 7 794
          NM_001018020.1 NP_001018020.1 8 954
          NM_001301244.1 NP_001288173.1 8 954
          NM_001301289.1 NP_001288218.1 7 794
          XM_005254637.1 XP_005254694.1 9 1035
          XM_005254638.3 XP_005254695.2 8 1397
          XM_005254639.3 XP_005254696.2 8 1397
          XM_005254645.2 XP_005254702.2 8 1397
          XM_005254647.3 XP_005254704.1 7 780
          XM_005254650.3 XP_005254707.1 7 780
          XM_005254651.1 XP_005254708.1 7 780
          XM_005254652.1 XP_005254709.1 7 780
          XM_005254653.2 XP_005254710.1 7 780
          XM_006720667.3 XP_006720730.2 8 1397
          XM_017022538.1 XP_016878027.1 8 1397
          XM_017022539.1 XP_016878028.1 8 1397
          XM_017022541.1 XP_016878030.1 7 780
          GenBank mRNA AB209041.1 - 7 831
          AF474156.1 - 7 556
          AJ000147.1 - 1 55
          AJ001055.1 - 1 55
          AK057989.1 - 10 740
          AK092051.1 - 6 2612
          AK092573.1 - 7 1852
          AK129882.1 - 7 641
          AK131384.1 - 9 968
          AK299387.1 - 8 843
          AK301319.1 - 7 690
          AL050179.1 - 8 835
          AY640414.1 - 8 763
          AY640415.1 - 8 763
          BC007433.2 - 8 904 2962925
          BC050473.1 - 7 781
          BC053545.1 - 7 792 5730451
          BX458841.2 - 8 823
          CD679271.1 - 4 285
          DQ424903.1 - 7 655
          GQ472198.1 - 8 904
          KU178366.1 - 8 763
          KU178367.1 - 8 763
          M19713.1 - 8 819
          M19714.1 - 8 705
          M19715.1 - 7 600
          X12369.1 - 7 593
          Z24727.1 - 978

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001301336, NM_001301736, NM_001034070, NM_001034068, NM_019131, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.

          Matching Taqman Gene Expression Assay:


          Hs00165966_m1

          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of heart rate by epinephrine
          movement of cell or subcellular component
          muscle contraction
          regulation of muscle contraction
          cytoskeleton organization
          regulation of heart contraction
          regulation of cell shape
          muscle filament sliding
          negative regulation of cell migration
          ruffle organization
          positive regulation of ATPase activity
          cellular response to reactive oxygen species
          wound healing
          sarcomere organization
          positive regulation of cell adhesion
          positive regulation of stress fiber assembly
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          negative regulation of vascular smooth muscle cell proliferation
          negative regulation of vascular associated smooth muscle cell migration
          actin binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          structural constituent of muscle

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0