Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
TPM1
Gene Name:
tropomyosin 1 (alpha)
Gene Aliases:
C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
Chromosome Location:
Chr.15: 63042196 - 63071915 on Build GRCh38
UniGene ID:
Hs.133892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14342
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000366.5
NP_000357.3
7
850
NM_001018004.1
NP_001018004.1
7
850
NM_001018005.1
NP_001018005.1
7
850
NM_001018006.1
NP_001018006.1
7
850
NM_001018007.1
NP_001018007.1
7
850
NM_001018008.1
NP_001018008.1
6
690
NM_001018020.1
NP_001018020.1
7
850
NM_001301244.1
NP_001288173.1
7
850
NM_001301289.1
NP_001288218.1
6
690
XM_005254637.1
XP_005254694.1
8
931
XM_005254638.3
XP_005254695.2
7
1293
XM_005254639.3
XP_005254696.2
7
1293
XM_005254645.2
XP_005254702.2
7
1293
XM_005254646.2
XP_005254703.1
6
676
XM_005254647.3
XP_005254704.1
6
676
XM_005254650.3
XP_005254707.1
6
676
XM_005254651.1
XP_005254708.1
6
676
XM_005254652.1
XP_005254709.1
6
676
XM_005254653.2
XP_005254710.1
6
676
XM_006720667.3
XP_006720730.2
7
1293
XM_017022534.1
XP_016878023.1
7
1293
XM_017022535.1
XP_016878024.1
7
805
XM_017022536.1
XP_016878025.1
7
1293
XM_017022537.1
XP_016878026.1
7
805
XM_017022538.1
XP_016878027.1
7
1293
XM_017022539.1
XP_016878028.1
7
1293
XM_017022540.1
XP_016878029.1
6
676
XM_017022541.1
XP_016878030.1
6
676
GenBank mRNA
AB209041.1
-
6
727
AF474156.1
-
6
452
AK057989.1
-
9
636
AK092051.1
-
5
2508
AK129882.1
-
6
537
AK131384.1
-
8
864
AK298625.1
-
6
566
AK299387.1
-
7
739
AK301319.1
-
6
586
AL050179.1
-
7
731
AY640414.1
-
7
659
AY640415.1
-
7
659
BC007433.2
-
7
800
2962925
BC050473.1
-
6
677
BC053545.1
-
6
688
5730451
BX458841.2
-
7
719
CD679271.1
-
3
181
DQ424903.1
-
6
551
GQ472198.1
-
7
800
KU178366.1
-
7
659
KU178367.1
-
7
659
M19713.1
-
7
715
M19714.1
-
7
601
M19715.1
-
6
496
X12369.1
-
6
489
Z24727.1
-
874
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00165966_m1
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
movement of cell or subcellular component
muscle contraction
regulation of muscle contraction
cytoskeleton organization
regulation of heart contraction
regulation of cell shape
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
negative regulation of vascular smooth muscle cell proliferation
negative regulation of vascular associated smooth muscle cell migration
actin binding
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0