Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSP90B2P SIRNA Assays ›
          Gene Symbol:
          HSP90B2P HSP90B1
          Gene Name:
          heat shock protein 90 beta family member 2, pseudogene
          heat shock protein 90 beta family member 1
          Gene Aliases:
          GRP94P1, GRP94b, HSP, HSPCP2, TRA1P1, TRAP1
          ECGP, GP96, GRP94, HEL-S-125m, HEL35, TRA1
          Chromosome Location:
          Chr.15: 99257525 - 99260276 on Build GRCh38
          Chromosome Location:
          Chr.12: 103930334 - 103947930 on Build GRCh38
          UniGene ID:
          Hs.656611
          Hs.192374
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s14374

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSP90B2P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_073383.1 1 1590
          GenBank mRNA AY956768.1 - 1 471
          Gene Symbol: HSP90B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003299.2 NP_003290.1 12 1682
          GenBank mRNA AB209534.1 - 12 1578
          AF087988.1 - 5 486
          AJ890084.1 - 12 1634
          AK025459.1 - 12 1634
          AK295262.1 - 6 604
          AK300521.1 - 8 855
          AY040226.1 - 11 1437
          BC066656.1 - 12 1596 6165138
          EU794608.1 - 12 1596
          FJ460515.1 - 12 1596
          X15187.1 - 12 1605

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          Hsp90 family chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          response to stress
          response to hypoxia
          toll-like receptor signaling pathway
          receptor-mediated endocytosis
          protein transport
          ER-associated ubiquitin-dependent protein catabolic process
          retrograde protein transport, ER to cytosol
          actin rod assembly
          protein folding in endoplasmic reticulum
          response to endoplasmic reticulum stress
          ATF6-mediated unfolded protein response
          negative regulation of apoptotic process
          regulation of phosphoprotein phosphatase activity
          sequestering of calcium ion
          cellular response to ATP
          ATP binding
          unfolded protein binding
          RNA binding
          calcium ion binding
          protein binding
          protein phosphatase binding
          virion binding
          low-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0