Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BIRC2 SIRNA Assays ›
          Gene Symbol:
          BIRC2
          Gene Name:
          baculoviral IAP repeat containing 2
          Gene Aliases:
          API1, HIAP2, Hiap-2, MIHB, RNF48, c-IAP1, cIAP1
          Chromosome Location:
          Chr.11: 102347182 - 102378670 on Build GRCh38
          UniGene ID:
          Hs.696238
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1449
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BIRC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166.4 NP_001157.1 2 1498
          NM_001256163.1 NP_001243092.1 2 1775
          GenBank mRNA AK225856.1 - 1 490
          AK313446.1 - 1 303
          BC016174.1 - 1 74 3908352
          BC028578.1 - 2 1776 4831062
          BX647978.1 - 2 1445
          L49431.1 - 1 240
          U37547.1 - 1 1205
          U45879.1 - 1 283

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00236911_m1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          response to hypoxia
          placenta development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular component disassembly involved in execution phase of apoptosis
          cell surface receptor signaling pathway
          I-kappaB kinase/NF-kappaB signaling
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          tumor necrosis factor-mediated signaling pathway
          regulation of toll-like receptor signaling pathway
          TRIF-dependent toll-like receptor signaling pathway
          NIK/NF-kappaB signaling
          regulation of RIG-I signaling pathway
          regulation of cell proliferation
          regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of innate immune response
          response to ethanol
          regulation of cell differentiation
          regulation of inflammatory response
          protein heterooligomerization
          response to cAMP
          regulation of cell cycle
          regulation of necroptotic process
          necroptotic process
          regulation of nucleotide-binding oligomerization domain containing signaling pathway
          mitotic spindle assembly
          negative regulation of ripoptosome assembly involved in necroptotic process
          positive regulation of protein K63-linked ubiquitination
          positive regulation of protein K48-linked ubiquitination
          positive regulation of protein monoubiquitination
          inhibition of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of cysteine-type endopeptidase activity
          regulation of reactive oxygen species metabolic process
          transcription coactivator activity
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          transferase activity
          ligase activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0