Search
Search

Ccdc68| Gene Symbol: Ccdc68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077667.1 | NP_001071135.1 | 3 | 306 | ||
| XM_008772113.1 | XP_008770335.1 | 4 | 600 | |||
| XM_008772114.1 | XP_008770336.1 | 3 | 756 | |||
| XM_008772115.1 | XP_008770337.1 | 3 | 379 | |||
| XM_008772116.1 | XP_008770338.1 | 3 | 385 | |||
| GenBank mRNA | BC127517.1 | - | 3 | 306 | 7104038 | |