Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOA1 SIRNA Assays ›
          Gene Symbol:
          APOA1
          Gene Name:
          apolipoprotein A1
          Gene Aliases:
          apo(a)
          Chromosome Location:
          Chr.11: 116835751 - 116837950 on Build GRCh38
          UniGene ID:
          Hs.93194
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1468

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APOA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000039.2 NP_000030.1 3 497
          NM_001318017.1 NP_001304946.1 3 210
          NM_001318018.1 NP_001304947.1 3 202
          NM_001318021.1 NP_001304950.1 3 453
          GenBank mRNA A14829.1 - 2 150
          A15879.1 - 1 59
          AK292231.1 - 3 171
          AL531353.3 - 3 202
          AL557908.3 - 3 210
          BC005380.1 - 3 169 3934992
          BC110286.1 - 3 192 5164525
          BU600916.1 - 3 128
          CD558050.1 - 3 497
          M11791.1 - 2 131
          M27875.1 - 2 150
          M29068.1 - 1 77
          T68127.1 - 3 210
          X00566.1 - 2 166
          X02162.1 - 3 217

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          regulation of protein phosphorylation
          endothelial cell proliferation
          platelet degranulation
          negative regulation of cytokine secretion involved in immune response
          phospholipid metabolic process
          phosphatidylcholine biosynthetic process
          cholesterol biosynthetic process
          receptor-mediated endocytosis
          transforming growth factor beta receptor signaling pathway
          G-protein coupled receptor signaling pathway
          integrin-mediated signaling pathway
          response to nutrient
          cholesterol metabolic process
          glucocorticoid metabolic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          negative regulation of very-low-density lipoprotein particle remodeling
          peripheral nervous system axon regeneration
          protein oxidation
          peptidyl-methionine modification
          triglyceride catabolic process
          lipid storage
          regulation of intestinal cholesterol absorption
          cholesterol transport
          adrenal gland development
          organ regeneration
          neuron projection regeneration
          regulation of Cdc42 protein signal transduction
          cholesterol efflux
          phospholipid efflux
          negative regulation of heterotypic cell-cell adhesion
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          high-density lipoprotein particle clearance
          positive regulation of Rho protein signal transduction
          lipoprotein metabolic process
          lipoprotein biosynthetic process
          response to drug
          cholesterol homeostasis
          blood vessel endothelial cell migration
          response to estrogen
          reverse cholesterol transport
          cellular protein metabolic process
          positive regulation of fatty acid biosynthetic process
          negative regulation of interleukin-1 beta secretion
          negative regulation of inflammatory response
          protein stabilization
          negative chemotaxis
          positive regulation of lipoprotein lipase activity
          vitamin transport
          positive regulation of hydrolase activity
          positive regulation of stress fiber assembly
          transmembrane transport
          phospholipid homeostasis
          negative regulation of lipase activity
          negative regulation of cell adhesion molecule production
          negative regulation of response to cytokine stimulus
          triglyceride homeostasis
          ERK1 and ERK2 cascade
          cholesterol import
          positive regulation of substrate adhesion-dependent cell spreading
          beta-amyloid binding
          protein binding
          phospholipid binding
          phospholipid transporter activity
          high-density lipoprotein particle binding
          cholesterol binding
          cholesterol transporter activity
          enzyme binding
          phosphatidylcholine binding
          apolipoprotein receptor binding
          apolipoprotein A-I receptor binding
          identical protein binding
          chemorepellent activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0