Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UROD SIRNA Assays ›
          Gene Symbol:
          UROD
          Gene Name:
          uroporphyrinogen decarboxylase
          Gene Aliases:
          PCT, UPD
          Chromosome Location:
          Chr.1: 45012133 - 45015669 on Build GRCh38
          UniGene ID:
          Hs.78601
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14712
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UROD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000374.4 NP_000365.3 3 335
          NR_036510.1 3 385
          XM_005271169.1 XP_005271226.1 3 290
          XM_005271170.1 XP_005271227.1 3 214
          GenBank mRNA AF104421.1 - 3 220
          AF104422.1 - 3 220
          AF104423.1 - 3 220
          AF104424.1 - 3 220
          AF104425.1 - 3 220
          AF104426.1 - 3 220
          AF104427.1 - 3 220
          AF104428.1 - 3 220
          AF104429.1 - 3 220
          AF104430.1 - 3 220
          AF104431.1 - 3 220
          AF104432.1 - 3 220
          AF104433.1 - 3 220
          AF104434.1 - 3 220
          AF104435.1 - 3 220
          AF104436.1 - 3 220
          AF104437.1 - 3 220
          AF104438.1 - 3 220
          AF104439.1 - 3 220
          AF104440.1 - 3 220
          AK291877.1 - 3 245
          AK293702.1 - 2 280
          AK295284.1 - 3 220
          AK309446.1 - 3 291
          AK310208.1 - 3 290
          AK315878.1 - 3 282
          BC001778.1 - 3 230 3542421
          BM554255.1 - 3 340
          BT006737.1 - 3 202
          BX647308.1 - 3 267
          CR456976.1 - 3 202
          CR542057.1 - 3 202
          DB462002.1 - 3 335
          M14016.1 - 3 220

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00165983_m1

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          liver development
          protoporphyrinogen IX biosynthetic process
          heme biosynthetic process
          response to iron ion
          response to amine
          response to estradiol
          response to ethanol
          uroporphyrinogen III metabolic process
          response to mercury ion
          response to methylmercury
          response to fungicide
          cellular response to arsenic-containing substance
          uroporphyrinogen decarboxylase activity
          protein binding
          ferrous iron binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0