Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NODAL SIRNA Assays ›
Gene Symbol:
Nodal
Gene Name:
nodal growth differentiation factor
Gene Aliases:
-
Chromosome Location:
Chr.20: 31035729 - 31044096 on Build Rnor_6.0
UniGene ID:
Rn.218528
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s148623
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nodal
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001106394.1
NP_001099864.1
2
620
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
trophectodermal cell differentiation
trophectodermal cellular morphogenesis
neural fold formation
liver development
placenta development
embryonic placenta development
maternal placenta development
vasculature development
heart looping
inhibition of neuroepithelial cell differentiation
determination of left/right symmetry
gastrulation
nervous system development
brain development
endoderm development
heart development
positive regulation of cell proliferation
embryonic pattern specification
anterior/posterior axis specification
anterior/posterior pattern specification
regulation of signal transduction
polarity specification of proximal/distal axis
regulation of gastrulation
positive regulation of vascular endothelial growth factor production
negative regulation of cell development
positive regulation of pathway-restricted SMAD protein phosphorylation
cell migration
stem cell population maintenance
positive regulation of cell-cell adhesion
lung development
positive regulation of activin receptor signaling pathway
floor plate morphogenesis
embryonic heart tube development
endodermal cell differentiation
nodal signaling pathway
growth
cell migration involved in gastrulation
regulation of apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of MAPK cascade
cell fate commitment
negative regulation of cell differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
axial mesodermal cell fate specification
mesendoderm development
cell development
digestive tract morphogenesis
anatomical structure formation involved in morphogenesis
embryonic cranial skeleton morphogenesis
tissue morphogenesis
formation of anatomical boundary
positive regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
digestive system development
embryonic process involved in female pregnancy
maternal process involved in parturition
positive regulation of SMAD protein import into nucleus
SMAD protein signal transduction
left lung morphogenesis
negative regulation of androgen receptor signaling pathway
epiblast cell-extraembryonic ectoderm cell signaling involved in anterior/posterior axis specification
positive regulation of ERK1 and ERK2 cascade
primitive streak formation
transforming growth factor beta receptor signaling pathway involved in primitive streak formation
nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
negative regulation of trophoblast cell migration
negative regulation of chorionic trophoblast cell proliferation
positive regulation of receptor activity
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
receptor agonist activity
type I activin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0