Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other YWHAZ SIRNA Assays ›
Gene Symbol:
YWHAZ
Gene Name:
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases:
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
Chromosome Location:
Chr.8: 100918576 - 100954068 on Build GRCh38
UniGene ID:
Hs.492407
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14971
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: YWHAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135699.1
NP_001129171.1
2
295
NM_001135700.1
NP_001129172.1
2
249
NM_001135701.1
NP_001129173.1
2
298
NM_001135702.1
NP_001129174.1
2
317
NM_003406.3
NP_003397.1
2
278
NM_145690.2
NP_663723.1
2
352
XM_005251061.3
XP_005251118.1
4
675
XM_005251063.3
XP_005251120.1
2
447
XM_017013810.1
XP_016869299.1
3
464
XM_017013811.1
XP_016869300.1
4
660
GenBank mRNA
AA830489.1
-
2
295
AK289945.1
-
2
188
AK307506.1
-
3
155
BC003623.2
-
2
245
2988020
BC013265.1
-
2
243
BC051814.1
-
2
366
BC063824.1
-
2
217
6180974
BC068456.1
-
2
266
4793291
BC072426.1
-
2
247
6315583
BC073141.1
-
2
201
BC083508.1
-
2
245
5563061
BC099904.1
-
2
267
5298373
BC101483.1
-
2
200
8068989
BC108281.1
-
2
255
3869494
BC111951.1
-
2
200
8327419
CN276627.1
-
2
243
CR996247.1
-
2
249
DB472116.1
-
2
334
DC298291.1
-
2
278
EU794582.1
-
2
246
EU794583.1
-
2
266
GQ472215.1
-
2
266
M86400.1
-
2
224
S79449.1
-
1
93
U28964.1
-
2
266
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00237047_m1
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
signal transduction
platelet activation
negative regulation of apoptotic process
regulation of mRNA stability
establishment of Golgi localization
membrane organization
Golgi reassembly
cell-cell adhesion
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
protein binding
transcription factor binding
protein kinase binding
protein domain specific binding
ubiquitin protein ligase binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0