Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFX SIRNA Assays ›
          Gene Symbol:
          ZFX
          Gene Name:
          zinc finger protein, X-linked
          Gene Aliases:
          ZNF926
          Chromosome Location:
          Chr.X: 24148994 - 24216255 on Build GRCh38
          UniGene ID:
          Hs.336681
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s14990

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178084.1 NP_001171555.1 8 1692
          NM_001178085.1 NP_001171556.1 11 2036
          NM_001178086.1 NP_001171557.1 6 1097
          NM_001178095.1 NP_001171566.1 8 1830
          NM_003410.3 NP_003401.2 10 1900
          XM_005274591.4 XP_005274648.1 9 2066
          XM_005274592.3 XP_005274649.1 8 1881
          XM_005274594.4 XP_005274651.1 7 3071
          XM_006724513.3 XP_006724576.1 11 2295
          XM_006724515.3 XP_006724578.1 10 2156
          XM_011545578.1 XP_011543880.1 11 2177
          XM_011545579.1 XP_011543881.1 10 1965
          XM_011545581.2 XP_011543883.1 10 2107
          XM_011545583.2 XP_011543885.1 7 1621
          XM_017029788.1 XP_016885277.1 11 2218
          XM_017029789.1 XP_016885278.1 10 2780
          XM_017029790.1 XP_016885279.1 10 2185
          XM_017029791.1 XP_016885280.1 9 2075
          XM_017029792.1 XP_016885281.1 9 1954
          XM_017029793.1 XP_016885282.1 8 1752
          XM_017029794.1 XP_016885283.1 10 2006
          XM_017029795.1 XP_016885284.1 7 1635
          XM_017029796.1 XP_016885285.1 8 1646
          XM_017029797.1 XP_016885286.1 11 2246
          XM_017029798.1 XP_016885287.1 7 1866
          XM_017029799.1 XP_016885288.1 7 1740
          XM_017029800.1 XP_016885289.1 6 1727
          XM_017029801.1 XP_016885290.1 6 1484
          GenBank mRNA AB209892.1 - 8 1863
          AF454950.1 - 4 807
          AY072770.1 - 3 654
          BC136308.1 - 10 1888 9020295
          BC136309.1 - 10 1888 9020296
          M30608.1 - 9 1851
          X59738.1 - 6 1094
          X59739.1 - 8 1668
          X59740.1 - 11 2017

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0