Search
Search

Serpina6| Gene Symbol: Serpina6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009663.1 | NP_001009663.1 | 2 | 571 | ||
| GenBank mRNA | BC088300.1 | - | 2 | 571 | 7323342 | |
| FQ209415.1 | - | 2 | 616 | |||
| FQ211289.1 | - | 2 | 586 | |||
| FQ218294.1 | - | 2 | 591 | |||
| FQ218398.1 | - | 2 | 580 | |||
| FQ219289.1 | - | 2 | 645 | |||