Search
Search

Nsmce2| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024876.1 | NP_001020047.1 | 4, 5 | 498 | ||
| XM_008765505.1 | XP_008763727.1 | 3, 4 | 2658 | |||
| GenBank mRNA | BC097477.1 | - | 4, 5 | 498 | 7458086 | |