Search
Search

Eda| Gene Symbol: Eda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257110.2 | XP_006257172.1 | 7 | 1101 | ||
| XM_006257111.2 | XP_006257173.1 | 7 | 1101 | |||
| XM_006257112.2 | XP_006257174.1 | 7 | 1092 | |||
| XM_228582.8 | XP_228582.6 | 7 | 1092 | |||