Search
Search

Rnase6
LOC103694852| Gene Symbol: Rnase6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206815.1 | NP_996538.1 | 1 | 179 | ||
| XM_006251871.2 | XP_006251933.1 | 2 | 248 | |||
| XM_006251872.2 | XP_006251934.1 | 2 | 547 | |||
| XM_008770517.1 | XP_008768739.1 | 3 | 392 | |||
| XM_008770518.1 | XP_008768740.1 | 3 | 366 | |||
| XM_008770519.1 | XP_008768741.1 | 2 | 266 | |||
| XM_008770520.1 | XP_008768742.1 | 2 | 245 | |||
| Gene Symbol: LOC103694852 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773787.1 | XP_008772009.1 | 2 | 248 | ||
| XM_008773788.1 | XP_008772010.1 | 2 | 547 | |||
| XM_008773789.1 | XP_008772011.1 | 3 | 395 | |||
| XM_008773790.1 | XP_008772012.1 | 3 | 369 | |||
| XM_008773791.1 | XP_008772013.1 | 2 | 269 | |||
| XM_008773792.1 | XP_008772014.1 | 2 | 245 | |||