Search
Search

Atp11a| Gene Symbol: Atp11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107324.2 | NP_001100794.2 | 17 | 2101 | ||
| XM_006253445.2 | XP_006253507.1 | 17 | 2056 | |||
| XM_006253446.2 | XP_006253508.1 | 17 | 2056 | |||
| XM_006253447.2 | XP_006253509.1 | 17 | 2056 | |||
| XM_006253448.2 | XP_006253510.1 | 17 | 2056 | |||