Search
Search

Xrcc4| Gene Symbol: Xrcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006999.1 | NP_001007000.1 | 3 | 284 | ||
| XM_006231751.1 | XP_006231813.1 | 3 | 453 | |||
| XM_008760636.1 | XP_008758858.1 | 3 | 415 | |||
| XM_008760637.1 | XP_008758859.1 | 3 | 480 | |||
| XM_008760638.1 | XP_008758860.1 | 3 | 415 | |||
| GenBank mRNA | BC083848.1 | - | 3 | 284 | 7122361 | |