Search
Search

Trim46| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107691.1 | NP_001101161.1 | 4 | 740 | ||
| XM_008761170.1 | XP_008759392.1 | 4 | 857 | |||
| XM_008761171.1 | XP_008759393.1 | 4 | 704 | |||
| XM_008761172.1 | XP_008759394.1 | 4 | 703 | |||
| XM_008761173.1 | XP_008759395.1 | 4 | 704 | |||