Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARNT SIRNA Assays ›
          Gene Symbol:
          ARNT
          Gene Name:
          aryl hydrocarbon receptor nuclear translocator
          Gene Aliases:
          HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
          Chromosome Location:
          Chr.1: 150809705 - 150876768 on Build GRCh38
          UniGene ID:
          Hs.632446
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s1613

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARNT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001197325.1 NP_001184254.1 6, 7 865
          NM_001286035.1 NP_001272964.1 8, 9 966
          NM_001286036.1 NP_001272965.1 7, 8 910
          NM_001668.3 NP_001659.1 7, 8 910
          NM_178427.2 NP_848514.1 6, 7 865
          XM_005245151.1 XP_005245208.1 7, 8 852
          XM_005245153.1 XP_005245210.1 7, 8 852
          XM_005245154.3 XP_005245211.1 8, 9 880
          XM_005245157.1 XP_005245214.1 7, 8 852
          XM_011509542.2 XP_011507844.1 7, 8 913
          XM_011509543.2 XP_011507845.1 8, 9 908
          XM_011509544.2 XP_011507846.1 7, 8 877
          XM_011509545.2 XP_011507847.1 6, 7 868
          XM_011509546.1 XP_011507848.1 6, 7 628
          XM_011509547.2 XP_011507849.1 6, 7 868
          XM_017001288.1 XP_016856777.1 8, 9 755
          XM_017001289.1 XP_016856778.1 8, 9 755
          XM_017001290.1 XP_016856779.1 7, 8 709
          XM_017001291.1 XP_016856780.1 7, 8 711
          XM_017001292.1 XP_016856781.1 8, 9 756
          XM_017001293.1 XP_016856782.1 7, 8 713
          XM_017001294.1 XP_016856783.1 7, 8 852
          XM_017001295.1 XP_016856784.1 7, 8 852
          XM_017001296.1 XP_016856785.1 6, 7 816
          GenBank mRNA AB209877.1 - 7, 8 721
          AF001307.1 - 7, 8 764
          AK223459.1 - 774
          AK290177.1 - 7, 8 849
          AK291705.1 - 6, 7 707
          AK293027.1 - 7, 8 910
          AK304772.1 - 6, 7 699
          AK313780.1 - 7, 8 851
          AK315812.1 - 6, 7 704
          AL834279.1 - 8, 9 905
          BC041121.1 - 7, 8 724
          BC060838.1 - 7, 8 845 5300334
          M69238.1 - 7, 8 765

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          embryonic placenta development
          positive regulation of endothelial cell proliferation
          transcription, DNA-templated
          positive regulation of vascular endothelial growth factor production
          cell differentiation
          intracellular receptor signaling pathway
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          positive regulation of protein sumoylation
          mRNA transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase II promoter in response to oxidative stress
          positive regulation of erythrocyte differentiation
          positive regulation of glycolytic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hormone biosynthetic process
          regulation of transcription from RNA polymerase II promoter in response to hypoxia
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          aryl hydrocarbon receptor activity
          protein binding
          transcription factor binding
          aryl hydrocarbon receptor binding
          enhancer binding
          sequence-specific DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0