Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPFIBP2 SIRNA Assays ›
Gene Symbol:
PPFIBP2
Gene Name:
PPFIA binding protein 2
Gene Aliases:
Cclp1
Chromosome Location:
Chr.11: 7513765 - 7667329 on Build GRCh38
UniGene ID:
Hs.655714
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16132
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPFIBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256568.1
NP_001243497.1
21
2273
NM_001256569.1
NP_001243498.1
20
2253
NM_003621.3
NP_003612.2
24
2850
XM_005253177.2
XP_005253234.1
24
2845
XM_006718349.2
XP_006718412.1
25
2878
XM_006718350.2
XP_006718413.1
25
2878
XM_006718351.2
XP_006718414.1
23
2686
XM_006718352.2
XP_006718415.1
22
2653
XM_006718353.2
XP_006718416.1
21
2245
XM_011520413.2
XP_011518715.1
26
2866
XM_011520414.2
XP_011518716.1
25
2593
XM_011520415.2
XP_011518717.1
25
2878
XM_011520416.2
XP_011518718.1
25
2878
XM_011520417.2
XP_011518719.1
23
2733
XM_011520418.2
XP_011518720.1
22
2330
XM_011520419.2
XP_011518721.1
23
2386
XM_011520420.2
XP_011518722.1
23
2329
XM_017018443.1
XP_016873932.1
25
2833
XM_017018444.1
XP_016873933.1
23
2686
XM_017018445.1
XP_016873934.1
22
2700
XM_017018446.1
XP_016873935.1
22
2653
XM_017018447.1
XP_016873936.1
21
3946
GenBank mRNA
AF034803.1
-
21
2178
AK001131.1
-
8
778
AK025337.1
-
20
2172
AK294554.1
-
13
1392
AK296718.1
-
21
2273
AK308398.1
-
24
2616
AK311048.1
-
14
1419
AL834426.1
-
23
2479
BC021026.1
-
7
821
BC021714.1
-
24
2798
4826610
CR749258.1
-
22
2287
Back To Top
More Information
Gene Ontology Categories:
Process(es)
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline