Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARNTL SIRNA Assays ›
Gene Symbol:
ARNTL
Gene Name:
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases:
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location:
Chr.11: 13276613 - 13387268 on Build GRCh38
UniGene ID:
Hs.65734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1616
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARNTL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001030272.2
NP_001025443.1
11
1176
NM_001030273.2
NP_001025444.1
12
1247
NM_001178.5
NP_001169.3
12
1231
NM_001297719.1
NP_001284648.1
12
1231
NM_001297722.1
NP_001284651.1
11
1176
NM_001297724.1
NP_001284653.1
12
1240
XM_006718233.3
XP_006718296.1
11
1118
XM_006718234.1
XP_006718297.1
12
1184
XM_011520105.2
XP_011518407.2
10
1282
XM_011520107.2
XP_011518409.2
10
1277
XM_011520108.1
XP_011518410.1
12
1224
XM_011520109.1
XP_011518411.1
10
979
XM_011520110.1
XP_011518412.1
13
1239
XM_011520111.1
XP_011518413.1
11
1054
XM_011520112.2
XP_011518414.1
13
1239
XM_011520113.1
XP_011518415.1
13
1239
XM_017017738.1
XP_016873227.1
10
1280
XM_017017739.1
XP_016873228.1
10
1276
XM_017017740.1
XP_016873229.1
11
1118
XM_017017741.1
XP_016873230.1
10
979
XM_017017742.1
XP_016873231.1
10
979
XM_017017743.1
XP_016873232.1
10
979
XM_017017744.1
XP_016873233.1
13
1239
XM_017017745.1
XP_016873234.1
12
1184
XM_017017746.1
XP_016873235.1
12
1194
XM_017017747.1
XP_016873236.1
11
1054
XM_017017748.1
XP_016873237.1
11
1111
GenBank mRNA
AB000812.1
-
9
812
AB000816.1
-
11
1223
AF044288.1
-
12
1141
AK095749.1
-
12
1132
AK291510.1
-
11
1089
AK315174.1
-
12
1169
BC016674.1
-
9
775
3921870
BC031214.1
-
12
1072
4793007
BC041129.1
-
12
1155
5753529
D89722.1
-
9
788
U51627.1
-
6
770
U60415.1
-
11
1075
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
circadian rhythm
negative regulation of TOR signaling
circadian regulation of gene expression
regulation of protein catabolic process
regulation of hair cycle
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of neurogenesis
regulation of insulin secretion
regulation of cell cycle
response to redox state
maternal process involved in parturition
positive regulation of canonical Wnt signaling pathway
oxidative stress-induced premature senescence
regulation of type B pancreatic cell development
negative regulation of glucocorticoid receptor signaling pathway
regulation of cellular senescence
positive regulation of skeletal muscle cell differentiation
transcription regulatory region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
core promoter sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II transcription factor binding
DNA binding
protein binding
aryl hydrocarbon receptor binding
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
Hsp90 protein binding
repressing transcription factor binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0