Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASPH SIRNA Assays ›
          Gene Symbol:
          Asph
          Gene Name:
          aspartate-beta-hydroxylase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 22577680 - 22799332 on Build Rnor_6.0
          UniGene ID:
          Rn.22695
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s162096

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Asph
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006237855.2 XP_006237917.1 2 662
          XM_006237858.2 XP_006237920.1 2 660
          XM_006237859.2 XP_006237921.1 2 1946
          XM_006237860.2 XP_006237922.1 2 1945
          XM_006237861.2 XP_006237923.1 2 662
          XM_006237862.2 XP_006237924.1 2 1947
          XM_006237863.2 XP_006237925.1 2 660
          XM_006237864.2 XP_006237926.1 2 1944
          XM_008763567.1 XP_008761789.1 2 662
          XM_008763568.1 XP_008761790.1 2 661
          XM_008763569.1 XP_008761791.1 2 663
          XM_008763570.1 XP_008761792.1 2 1947
          XM_008763571.1 XP_008761793.1 2 663
          XM_008763572.1 XP_008761794.1 2 661
          XM_008763573.1 XP_008761795.1 2 1946
          XM_008763574.1 XP_008761796.1 2 1947
          XM_008763575.1 XP_008761797.1 2 666
          GenBank mRNA BC085901.1 - 2 238 7193538

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          pattern specification process
          negative regulation of cell proliferation
          positive regulation of calcium ion transport into cytosol
          peptidyl-amino acid modification
          regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          regulation of protein stability
          activation of store-operated calcium channel activity
          response to ATP
          limb morphogenesis
          peptidyl-aspartic acid hydroxylation
          positive regulation of proteolysis
          positive regulation of transcription, DNA-templated
          regulation of cytosolic calcium ion concentration
          palate development
          face morphogenesis
          calcium ion transmembrane transport
          cellular response to calcium ion
          positive regulation of intracellular protein transport
          activation of cysteine-type endopeptidase activity
          regulation of protein depolymerization
          molecular_function
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0