Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CADPS SIRNA Assays ›
Gene Symbol:
CADPS
Gene Name:
calcium dependent secretion activator
Gene Aliases:
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location:
Chr.3: 62398346 - 62875389 on Build GRCh38
UniGene ID:
Hs.654933
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16393
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CADPS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003716.3
NP_003707.2
30
4303
NM_183393.2
NP_899630.1
27
4066
NM_183394.2
NP_899631.1
28
4186
XM_006713378.2
XP_006713441.2
28
4124
XM_011534177.1
XP_011532479.1
33
4532
XM_011534178.1
XP_011532480.1
32
4514
XM_011534180.1
XP_011532482.1
33
4484
XM_011534182.1
XP_011532484.1
32
4463
XM_011534183.1
XP_011532485.1
31
4445
XM_011534189.1
XP_011532491.1
31
4367
XM_011534191.2
XP_011532493.1
29
3973
XM_011534192.1
XP_011532494.1
32
4325
XM_011534193.1
XP_011532495.1
32
4313
XM_011534194.2
XP_011532496.1
31
3955
XM_011534195.1
XP_011532497.1
30
4298
XM_011534196.1
XP_011532498.1
31
4295
XM_011534197.2
XP_011532499.1
28
4229
XM_011534198.2
XP_011532500.1
28
3775
XM_011534199.2
XP_011532501.1
28
3772
XM_011534200.2
XP_011532502.1
27
3757
XM_011534203.1
XP_011532505.1
30
4280
XM_017007358.1
XP_016862847.1
33
4538
XM_017007359.1
XP_016862848.1
33
4526
XM_017007360.1
XP_016862849.1
32
4517
XM_017007361.1
XP_016862850.1
32
4469
XM_017007362.1
XP_016862851.1
32
4457
XM_017007363.1
XP_016862852.1
31
4448
XM_017007364.1
XP_016862853.1
31
4370
XM_017007365.1
XP_016862854.1
31
4358
XM_017007366.1
XP_016862855.1
30
4352
XM_017007367.1
XP_016862856.1
30
3991
XM_017007368.1
XP_016862857.1
30
3988
XM_017007369.1
XP_016862858.1
32
4325
XM_017007370.1
XP_016862859.1
31
4310
XM_017007371.1
XP_016862860.1
30
4301
XM_017007372.1
XP_016862861.1
31
3946
XM_017007373.1
XP_016862862.1
31
4298
XM_017007374.1
XP_016862863.1
30
4289
XM_017007375.1
XP_016862864.1
30
3931
XM_017007376.1
XP_016862865.1
29
4283
XM_017007377.1
XP_016862866.1
29
3922
XM_017007378.1
XP_016862867.1
29
3919
XM_017007379.1
XP_016862868.1
29
4271
XM_017007380.1
XP_016862869.1
30
4214
XM_017007381.1
XP_016862870.1
30
4211
XM_017007382.1
XP_016862871.1
29
3844
XM_017007383.1
XP_016862872.1
29
4193
XM_017007384.1
XP_016862873.1
30
4163
XM_017007385.1
XP_016862874.1
30
4151
XM_017007386.1
XP_016862875.1
29
4142
GenBank mRNA
AB032947.2
-
30
4293
AB593079.1
-
28
4229
AF458662.1
-
27
4016
AK127506.1
-
3
3196
AL833895.1
-
28
3483
BC015754.1
-
8
634
BC072684.1
-
7
584
BC131731.1
-
21
2261
40124561
BX647166.1
-
10
918
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
protein transport
vesicle organization
synaptic vesicle priming
catecholamine secretion
dense core granule exocytosis
protein binding
lipid binding
protein kinase binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0