Search
Search

Dst| Gene Symbol: Dst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244701.2 | XP_006244763.1 | 32 | 6988 | ||
| XM_006244702.2 | XP_006244764.1 | 32 | 6988 | |||
| XM_006244703.2 | XP_006244765.1 | 32 | 6988 | |||
| XM_006244704.2 | XP_006244766.1 | 32 | 6988 | |||
| XM_006244705.2 | XP_006244767.1 | 32 | 6988 | |||
| XM_006244706.2 | XP_006244768.1 | 32 | 6989 | |||
| XM_006244707.2 | XP_006244769.1 | 36 | 5607 | |||
| XM_006244708.2 | XP_006244770.1 | 32 | 6989 | |||
| XM_006244713.2 | XP_006244775.1 | 39 | 6260 | |||
| XM_006244714.2 | XP_006244776.1 | 36 | 5561 | |||
| XM_008766964.1 | XP_008765186.1 | 37 | 5643 | |||
| XM_008766965.1 | XP_008765187.1 | 26 | 7747 | |||