Search
Search

Tuba4a| Gene Symbol: Tuba4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007004.1 | NP_001007005.1 | 3, 4 | 411 | ||
| XM_006245227.2 | XP_006245289.1 | 3, 4 | 810 | |||
| GenBank mRNA | BC083726.1 | - | 3, 4 | 411 | 7191229 | |
| FQ212913.1 | - | 435 | ||||
| FQ212965.1 | - | 3, 4 | 435 | |||
| FQ213109.1 | - | 3, 4 | 435 | |||
| FQ213658.1 | - | 3, 4 | 435 | |||
| FQ213871.1 | - | 3, 4 | 435 | |||
| FQ215031.1 | - | 3, 4 | 435 | |||
| FQ215251.1 | - | 3, 4 | 435 | |||
| FQ215445.1 | - | 3, 4 | 435 | |||
| FQ215549.1 | - | 3, 4 | 435 | |||
| FQ218669.1 | - | 3, 4 | 435 | |||
| FQ225165.1 | - | 3, 4 | 435 | |||
| FQ225303.1 | - | 3, 4 | 435 | |||
| FQ225881.1 | - | 3, 4 | 435 | |||
| FQ225974.1 | - | 3, 4 | 435 | |||
| FQ226026.1 | - | 3, 4 | 432 | |||
| FQ230256.1 | - | 3, 4 | 435 | |||
| FQ230321.1 | - | 3, 4 | 435 | |||
| FQ231447.1 | - | 3, 4 | 428 | |||
| FQ232744.1 | - | 3, 4 | 435 | |||
| FQ232854.1 | - | 3, 4 | 432 | |||
| FQ234185.1 | - | 3, 4 | 435 | |||
| FQ234507.1 | - | 3, 4 | 435 | |||
| FQ235001.1 | - | 3, 4 | 435 | |||