Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLIT2 SIRNA Assays ›
Gene Symbol:
Slit2
Gene Name:
slit homolog 2 (Drosophila)
Gene Aliases:
-
Chromosome Location:
Chr.14: 66831848 - 67170361 on Build Rnor_6.0
UniGene ID:
Rn.146652
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s165611
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Slit2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022632.2
NP_072154.2
23
2982
XM_008770210.1
XP_008768432.1
20
2117
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin superfamily cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
metanephros development
ureteric bud development
in utero embryonic development
negative regulation of protein phosphorylation
cell migration involved in sprouting angiogenesis
negative regulation of leukocyte chemotaxis
chemotaxis
axonogenesis
axon guidance
brain development
motor neuron axon guidance
negative regulation of cell proliferation
negative regulation of lamellipodium assembly
negative regulation of endothelial cell migration
negative regulation of gene expression
negative regulation of smooth muscle cell migration
single organismal cell-cell adhesion
spinal cord development
olfactory bulb development
chemorepulsion involved in embryonic olfactory bulb interneuron precursor migration
chemorepulsion involved in postnatal olfactory bulb interneuron migration
corticospinal neuron axon guidance through spinal cord
telencephalon cell migration
cell differentiation
negative regulation of cell growth
negative regulation of cell migration
negative regulation of axon extension
negative regulation of actin filament polymerization
retinal ganglion cell axon guidance
response to nutrient levels
cellular response to hormone stimulus
dorsal/ventral axon guidance
Roundabout signaling pathway
follicle-stimulating hormone signaling pathway
positive regulation of apoptotic process
negative regulation of catalytic activity
negative regulation of vascular permeability
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
axon extension involved in axon guidance
negative regulation of inflammatory response
negative chemotaxis
induction of negative chemotaxis
negative regulation of small GTPase mediated signal transduction
response to cortisol
mammary gland duct morphogenesis
mammary duct terminal end bud growth
apoptotic process involved in luteolysis
negative regulation of chemokine-mediated signaling pathway
cellular response to heparin
negative regulation of smooth muscle cell chemotaxis
negative regulation of mononuclear cell migration
negative regulation of neutrophil chemotaxis
negative regulation of monocyte chemotaxis
negative regulation of retinal ganglion cell axon guidance
negative regulation of cellular response to growth factor stimulus
follicle-stimulating hormone receptor activity
GTPase inhibitor activity
calcium ion binding
protein binding
heparin binding
identical protein binding
protein homodimerization activity
laminin-1 binding
proteoglycan binding
heparan sulfate proteoglycan binding
chemorepellent activity
Roundabout binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0