Search
Search

Cog3| Gene Symbol: Cog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012157.1 | NP_001012157.1 | 16 | 1867 | ||
| XM_006252323.2 | XP_006252385.1 | 16 | 2460 | |||
| GenBank mRNA | BC083901.1 | - | 16 | 1867 | 7133573 | |
| BC091298.1 | - | 1 | 75 | |||