Search
Search

Fgd3| Gene Symbol: Fgd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108409.1 | NP_001101879.1 | 11 | 1544 | ||
| XM_008771543.1 | XP_008769765.1 | 11 | 2095 | |||
| XM_008771544.1 | XP_008769766.1 | 11 | 2703 | |||
| XM_008771545.1 | XP_008769767.1 | 11 | 1797 | |||
| XM_008771546.1 | XP_008769768.1 | 11 | 1766 | |||