Search
Search

Gramd1a| Gene Symbol: Gramd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014160.1 | NP_001014182.1 | 3 | 272 | ||
| XM_006228832.2 | XP_006228894.1 | 5 | 687 | |||
| XM_006228833.2 | XP_006228895.1 | 5 | 685 | |||
| XM_006228834.2 | XP_006228896.1 | 4 | 453 | |||
| XM_006228835.2 | XP_006228897.1 | 4 | 451 | |||
| GenBank mRNA | AY171575.1 | - | 3 | 272 | ||
| BC105896.1 | - | 4 | 373 | 7380410 | ||