Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CFLAR SIRNA Assays ›
Gene Symbol:
CFLAR
Gene Name:
CASP8 and FADD like apoptosis regulator
Gene Aliases:
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
Chromosome Location:
Chr.2: 201116015 - 201172688 on Build GRCh38
UniGene ID:
Hs.390736
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16865
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CFLAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127183.2
NP_001120655.1
2
765
NM_001127184.2
NP_001120656.1
2
737
NM_001202516.1
NP_001189445.1
2
737
NM_001308042.1
NP_001294971.1
1
409
NM_001308043.1
NP_001294972.1
2
787
NM_003879.5
NP_003870.4
2
737
XM_017005189.1
XP_016860678.1
2
728
XM_017005190.1
XP_016860679.1
2
601
XM_017005191.1
XP_016860680.1
2
680
XM_017005192.1
XP_016860681.1
2
728
XM_017005193.1
XP_016860682.1
2
2745
XM_017005194.1
XP_016860683.1
2
601
XM_017005195.1
XP_016860684.1
2
718
XM_017005197.1
XP_016860686.1
2
2745
XM_017005198.1
XP_016860687.1
2
601
GenBank mRNA
AB209600.1
-
2
484
AF005774.1
-
2
707
AF005775.1
-
1
272
AF009616.1
-
2
684
AF009617.1
-
1
272
AF009618.1
-
1
272
AF009619.1
-
1
272
AF010127.1
-
2
682
AF015450.1
-
2
776
AF015451.1
-
1
272
AF015452.1
-
1
272
AF041458.1
-
1
287
AF041460.1
-
1
272
AF041461.1
-
1
227
AK289913.1
-
2
561
AK297387.1
-
3
634
AK308767.1
-
2
647
AK315208.1
-
2
508
BC001602.1
-
2
531
3542993
BT006751.1
-
1
272
U85059.1
-
1
272
U97074.1
-
2
654
U97075.1
-
1
565
Y14039.1
-
2
753
Y14040.1
-
2
753
Back To Top
More Information
Panther Classification:
Molecular Function -
protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
apoptotic process
skeletal muscle tissue development
skeletal muscle atrophy
regulation of skeletal muscle satellite cell proliferation
skeletal myofibril assembly
viral process
negative regulation of apoptotic process
positive regulation of catalytic activity
positive regulation of I-kappaB kinase/NF-kappaB signaling
skeletal muscle tissue regeneration
positive regulation of NF-kappaB transcription factor activity
regulation of necroptotic process
execution phase of apoptosis
negative regulation of myoblast fusion
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway
protease binding
death receptor binding
protein binding
enzyme activator activity
protein complex binding
cysteine-type endopeptidase activity involved in execution phase of apoptosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0