Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC100910882 SIRNA Assays ›
          Gene Symbol:
          LOC100910882 Rbm39
          Gene Name:
          RNA-binding protein 39-like
          RNA binding motif protein 39
          Gene Aliases:
          -
          Rnp1, Rnpc2, Rrm
          Chromosome Location:
          Chr.3: 152226018 - 152259228 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 151691505 - 151724715 on Build Rnor_6.0
          UniGene ID:
          Rn.8555
          Rn.8555
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s168782

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC100910882
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003749605.3 XP_003749653.1 7, 8 853
          XM_003749606.3 XP_003749654.1 7, 8 853
          XM_006235443.2 XP_006235505.1 7, 8 874
          XM_006235446.2 XP_006235508.1 7, 8 2295
          XM_008762415.1 XP_008760637.1 6, 7 790
          XM_008762416.1 XP_008760638.1 6, 7 811
          XM_008762417.1 XP_008760639.1 6, 7 6693
          XM_008762418.1 XP_008760640.1 6, 7 3785
          XM_008762419.1 XP_008760641.1 6, 7 5427
          XM_008762420.1 XP_008760642.1 6, 7 6714
          XM_008762421.1 XP_008760643.1 7, 8 5203
          XM_008762422.1 XP_008760644.1 6, 7 790
          Gene Symbol: Rbm39
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013207.2 NP_001013225.1 795
          XM_006235331.2 XP_006235393.1 853
          XM_006235332.2 XP_006235394.1 874
          XM_006235335.2 XP_006235397.1 2295
          XM_008762349.1 XP_008760571.1 790
          XM_008762350.1 XP_008760572.1 811
          XM_008762351.1 XP_008760573.1 6, 7 6693
          XM_008762352.1 XP_008760574.1 6, 7 3785
          XM_008762353.1 XP_008760575.1 5427
          XM_008762354.1 XP_008760576.1 6, 7 6714
          XM_008762355.1 XP_008760577.1 5203
          XM_008762356.1 XP_008760578.1 790
          GenBank mRNA BC078917.1 - 868
          BC091394.1 - 784 7322533

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_004902, NM_001242599, NM_184234, NM_001242600, L10910, and others.

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          nucleotide binding
          RNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0