Search
Search

Rbm39
LOC100910882| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013207.2 | NP_001013225.1 | 1201 | |||
| XM_006235331.2 | XP_006235393.1 | 1259 | ||||
| XM_006235332.2 | XP_006235394.1 | 1280 | ||||
| XM_006235335.2 | XP_006235397.1 | 2701 | ||||
| XM_008762349.1 | XP_008760571.1 | 1196 | ||||
| XM_008762350.1 | XP_008760572.1 | 1217 | ||||
| XM_008762351.1 | XP_008760573.1 | 10 | 7099 | |||
| XM_008762352.1 | XP_008760574.1 | 10 | 4191 | |||
| XM_008762353.1 | XP_008760575.1 | 5833 | ||||
| XM_008762354.1 | XP_008760576.1 | 10 | 7120 | |||
| XM_008762355.1 | XP_008760577.1 | 5609 | ||||
| XM_008762356.1 | XP_008760578.1 | 1196 | ||||
| GenBank mRNA | AY769432.1 | - | 53 | |||
| BC078917.1 | - | 1274 | ||||
| BC091394.1 | - | 1190 | 7322533 | |||
| Gene Symbol: LOC100910882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749605.3 | XP_003749653.1 | 11 | 1259 | ||
| XM_003749606.3 | XP_003749654.1 | 11 | 1259 | |||
| XM_006235443.2 | XP_006235505.1 | 11 | 1280 | |||
| XM_006235446.2 | XP_006235508.1 | 11 | 2701 | |||
| XM_008762415.1 | XP_008760637.1 | 10 | 1196 | |||
| XM_008762416.1 | XP_008760638.1 | 10 | 1217 | |||
| XM_008762417.1 | XP_008760639.1 | 10 | 7099 | |||
| XM_008762418.1 | XP_008760640.1 | 10 | 4191 | |||
| XM_008762419.1 | XP_008760641.1 | 10 | 5833 | |||
| XM_008762420.1 | XP_008760642.1 | 10 | 7120 | |||
| XM_008762421.1 | XP_008760643.1 | 11 | 5609 | |||
| XM_008762422.1 | XP_008760644.1 | 10 | 1196 | |||