Search
Search

Fam76a
LOC103690073| Gene Symbol: Fam76a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108686.1 | NP_001102156.1 | 3 | 374 | ||
| GenBank mRNA | FQ211538.1 | - | 2 | 394 | ||
| FQ222884.1 | - | 3 | 376 | |||
| Gene Symbol: LOC103690073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773876.1 | XP_008772098.1 | 455 | |||